Sample bag, secure-T
Sample bag, clear, with tear-off protection strip, Secure -T, 305×178 mm, 76 µm
LABPSCL7012
: LABPLAS
Secure-T® blender bags are are made to handle the most robust blending applications. They are available for all sizes of blenders and the tear-off top ensures internal sterility right up until the time of use, eliminating the risk of internal contamination.
- Compact and easy to use
- Open bags are gamma irradiated
- Maximum bag mouth opening to facilitate insertion of sample
- Description:Sample bag, clear, with tear-off protection strip, Secure -T
- L×W:305×178 mm
- Seq:01
- Thickness:76 µm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



