Order Entry
Denmark
ContactUsLinkComponent
Beakers, tall form
Beakers, tall form
Catalog # WITG5.504.150
Supplier:  witeg Labortechnik
undefined
Beakers, tall form
Catalog # WITG5.504.150
Supplier:  witeg Labortechnik

Specifications

  • Ext.Ø:
    53 mm
  • Capacity:
    150 ml
  • Height:
    96 mm
  • Pk:
    10

Specifications

About this item

Borosilicate glass 3.3 without spout.

  • Excellent chemical resistance
  • High temperature resistance
  • Autoclavable

Ideal for general laboratory use. With white graduations.