Order Entry
Denmark
ContactUsLinkComponent
 
Bottles, wide neck, with screw cap
  LPIT294221
Bottles, wide neck, with screw cap
  LPIT294221

 

  • Ext.Ø:
    46 mm
  • Capacity:
    100 ml
  • Height:
    93 mm
  • Neck-Ø:
    24 mm
  • Pk:
    140

 

 

LDPE, grey.

  • Good chemical resistance
  • Good temperature resistance

Screw cap with gasket.