Order Entry
Denmark
Orders LinkContactUsLinkComponent
Powder funnels, PP
Powder funnels, PP
Supplier:   Sampling Systems
Powder funnels, PP
Supplier:  Sampling Systems
Certificates
Powder funnels, PP
Powder funnels, PP
4 Options Available Below
Product Details & Documents
Reusable powder funnel, made of PP, crevice free, with wide diameter discharge spout to enable powder to be discharged without catching.
  • Good chemical resistance
  • Good temperature resistance
  • Ideal for loading powder into drums, containers or kegs
Product Family Options
Product Information
  • Funnel O-Ø
    Stem O-Ø
    Availability
    Price
  • 100 mm
    26 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 26,70 kr
    26,70 kr
    1 * 1 Items
       26,70 kr
  • 150 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 50,10 kr
    50,10 kr
    1 * 1 Items
       50,10 kr
  • 250 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 386,00 kr
    386,00 kr
    1 * 1 Items
       386,00 kr
  • 350 mm
    52 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 511,00 kr
    511,00 kr
    1 * 1 Items
       511,00 kr