Cyclopentyl methyl ether (CPME) ≥99.9% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:5614-37-9
:
Formula:C₆H₁₂O
MDL Number:MFCD08276401
:100.16 g/mol
Melting Pt:–140 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:–1 °C (closed cup)
UN:3271
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 50 ppm
:GHS02
:GHS07
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Colouration:Max. 10 APHA
- Peroxides (as H2O2):Max. 50 ppm
- Water:Max. 100 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
spec_VWRC84565.290 spec_VWRC84565.290 spec_VWRC84565.360 spec_VWRC84565.360
Product specification
VWR® Green Solvents for Sustainable Chemistry.pdf
Product Literature
Frequently Bought Together