Order Entry
Denmark
Orders LinkContactUsLinkComponent
Immersion coolers
Immersion coolers
Supplier:   witeg Labortechnik
Immersion coolers
Supplier:  witeg Labortechnik
Certificates
Immersion coolers
Immersion coolers
3 Options Available Below
Product Details & Documents
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For immersion into distillation assemblies etc. Can be used to regulate refluxing by adjusting the flow of air or water through it. Water connection with two glass olives.
Product Family Options
Product Information
  • Jacket length
    Cone size
    Availability
    Price
  • 100 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 644,00 kr
    644,00 kr
    1 * 1 Items
       644,00 kr
  • 100 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 625,00 kr
    625,00 kr
    1 * 1 Items
       625,00 kr
  • 140 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 823,00 kr
    823,00 kr
    1 * 1 Items
       823,00 kr