Order Entry
Denmark
Orders LinkContactUsLinkComponent
Distilling bridges
Distilling bridges
Supplier:   witeg Labortechnik
Distilling bridges
Supplier:  witeg Labortechnik
Certificates
Distilling bridges
Distilling bridges
4 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.
Product Family Options
Product Information
  • Jacket length
    Cone size
    Availability
    Price
  • 150 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.290,00 kr
    1.290,00 kr
    1 * 1 Items
       1.290,00 kr
  • 200 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.290,00 kr
    1.290,00 kr
    1 * 1 Items
       1.290,00 kr
  • 300 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.480,00 kr
    1.480,00 kr
    1 * 1 Items
       1.480,00 kr
  • 300 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.480,00 kr
    1.480,00 kr
    1 * 1 Items
       1.480,00 kr