Order Entry
Denmark
Orders LinkContactUsLinkComponent
Distilling heads
Distilling heads
Supplier:   witeg Labortechnik
Distilling heads
Supplier:  witeg Labortechnik
Certificates
Distilling heads
Distilling heads
4 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With one NS socket and two NS cones, descending angle: 75°.
Product Family Options
Product Information
  • Cone to condenser
    Cone to flask
    Availability
    Price
  • 19/26 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 412,00 kr
    412,00 kr
    1 * 1 Items
       412,00 kr
  • 14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 287,00 kr
    287,00 kr
    1 * 1 Items
       287,00 kr
  • 19/26 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 393,00 kr
    393,00 kr
    1 * 1 Items
       393,00 kr
  • 24/29 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 412,00 kr
    412,00 kr
    1 * 1 Items
       412,00 kr