Order Entry
Denmark
ContactUsLinkComponent
 
n-Dodecyl-β-D-maltoside ≥99.0%, high purity
Catalog # J424-1G
Supplier:  VWR Chemicals
CAS Number:  
undefined
n-Dodecyl-β-D-maltoside ≥99.0%, high purity
Catalog # J424-1G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Abs.@225 nm (1%, Water):
    0.1
  • Abs.@260 nm (1%, Water):
    0.06
  • Abs.@280 nm (1%, Water):
    0.04
  • Abs.@340 nm (1%, Water):
    0.02
  • Alpha (HPLC):
    15 %
  • pH (1%, Water) @25C:
    5 - 8
  • Purity:
    99.0 %
  • Solubility (1%, Water):
    PASS

Specification Test Results

About this item

Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
 :
 :510.6 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:69227-93-6
Formula:C₂₄H₄₆O₁₁
MDL Number:MFCD00043012