L-Lactate dehydrogenase (from Muscle)
Catalog # 427217-25
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:25 KU
- CAS No.:9001-60-9
- Source:Muscle
- Storage conditions:Refrigerator
- MDL No.:MFCD00131466
- Enzyme name:L-Lactate dehydrogenase
- Enzyme synonyms:LDH