Actinomycin D 10 mmol/l in aqueous solution
Catalog # API-01
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:50 µl
- Seq:1202
Specifications
About this item
:
:1255.5 g/mol
UN:3462
ADR:6.1,III
CAS Number:50-76-0
Formula:C₆₂H₈₆N₁₂O₁₆
MDL Number:MFCD00005033