Actinomycin D ≥98% (by HPLC)
Catalog # 40038.
Supplier: Biotium
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:1255.5 g/mol
UN:3462
ADR:6.1,II
CAS Number:50-76-0
Formula:C₆₂H₈₆N₁₂O₁₆
MDL Number:MFCD00005033