Actinomycin D 93%
Leverandør: Thermo Fisher Scientific
Certifikater
Om denne vare
CAS nummer:50-76-0
:
Formel:C₆₂H₈₆N₁₂O₁₆
MDL nummer:MFCD00005033
:1255.5 g/mol
Smeltende Pt:245…252 °C
Opbevaringstemperatur:Freezer
FN:3462
ADR:6.1,II
Specifikationstestresultater
- Appearance:Red Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥91 %
Specifikationstestresultater
Frequently Bought Together