Organic reference standards, amphotericin b, Ph. Eur.
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:1397-89-3
- Formula:C₄₇H₇₃NO₁₇
- Description:Amphotericin B for microbiological assay
Specifications
Frequently Bought Together