Order Entry
Denmark
Orders LinkContactUsLinkComponent
Malt extract

Malt extract

500 g
Catalog # OXOILP0039B
Malt extract
Catalog # OXOILP0039B
CAS Number:  8002-48-0
Malt extract

500 g

CAS Number:  8002-48-0
Price
523,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:8002-48-0
  • MDL Number:MFCD00131570
  • Storage Temperature:Ambient
  • EINECS:232-310-9

Documents

Specifications

Specifications

  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.