Order Entry
Denmark
ContactUsLinkComponent
Hexamethylphosphoramide-[D₁₈] (99.5% D) for NMR spectroscopy
Hexamethylphosphoramide-[D₁₈] (99.5% D) for NMR spectroscopy
Catalog # 8.15042.0005
Supplier:  Merck
CAS Number:  
Hexamethylphosphoramide-[D₁₈] (99.5% D) for NMR spectroscopy
Catalog # 8.15042.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
EINECS:257-063-4
CAS Number:51219-90-0
Formula:C₆D₁₈N₃OP