Chromotrope 2R, pure
:4197-07-3
:
:C₁₆H₁₀N₂Na₂O₈S₂
:MFCD00003955
:468.37 g/mol
:224-085-0
:GHS07
:Ambient
- Appearance:Red-brown Crystalline powder
- Infrared spectrum:Conforms
- Specific extinction E (1%/1cm):≥360 (509 nm, water)
Frequently Bought Together