Order Entry
Denmark
Orders LinkContactUsLinkComponent
Naphthol AS-MX phosphate

Naphthol AS-MX phosphate

Glass bottle for solids, 250 mg
Catalog # SERA30002.01
Irritant
Marketsource
Supplier:   Serva
Naphthol AS-MX phosphate
Catalog # SERA30002.01
Supplier:  Serva
Marketsource
CAS Number:  1596-56-1
Naphthol AS-MX phosphate

Glass bottle for solids, 250 mg

CAS Number:  1596-56-1
Price
313,64 kr
Log in for availability
1 * 250 mg
Product Details & Documents
  • CAS Number:1596-56-1
  • Formula:C₁₉H₁₈NO₅P
  • MDL Number:MFCD00042711
  • Storage Temperature:Freezer
  • EINECS:216-480-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg

Specifications