Order Entry
Denmark
ContactUsLinkComponent
 
 
 
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    241.33 g/mol
  • Boiling Point:
    168 - 170 °C
  • Flash Point:
    52 °C
  • Refractive Index:
    1.4018 (20 °C, 589 nm)
  • Density:
    1.03 g/cm3 (20 °C)

 

 

Frequently Bought Together