Order Entry
Denmark
ContactUsLinkComponent
 
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
Catalog # HYPETS-48920
CAS Number:  
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
Catalog # HYPETS-48920
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :241.33 g/mol
CAS Number:77377-52-7
Formula:C₉H₁₈F₃NOSi
MDL Number:MFCD00009671