Tris acetate
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
- CAS Number:6850-28-8
- Formula:NH₂C(CH₂OH)₃·CH₃COOH
- MDL Number:MFCD00038951
- Molecular Weight:181.1870 g/mol
- Storage Temperature:Ambient
- EINECS:229-939-6
2 Options Available Below
Product Details & Documents
Synonyms: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
- CAS Number:6850-28-8
- Formula:NH₂C(CH₂OH)₃·CH₃COOH
- MDL Number:MFCD00038951
- Molecular Weight:181.1870 g/mol
- Storage Temperature:Ambient
- EINECS:229-939-6
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



