Order Entry
Denmark
ContactUsLinkComponent
(±)-2-Methylglutaric acid for synthesis, Sigma-Aldrich®
(±)-2-Methylglutaric acid for synthesis, Sigma-Aldrich®
Catalog # 8.41328.0005
Supplier:  Merck
CAS Number:  
(±)-2-Methylglutaric acid for synthesis, Sigma-Aldrich®
Catalog # 8.41328.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Melting Point:
    74 - 76 °C
  • Molar Mass:
    146.14 g/mol
  • Synonyms:
    2-Methylpentanedioic acid
  • Hill Formula:
    C6H10O4

Specification Test Results

About this item

Synonyms: 2-Methylpentanedioic acid
 :
 :146.14 g/mol
EINECS:210-521-7
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:617-62-9
Formula:C₆H₁₀O₄
MDL Number:MFCD00002661
 :M-Clarity™ quality level = MQ200