(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:369-07-3
:
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00063255
:301.25 g/mol
Melting Pt:∼ 192 °C (dec.)
Storage Temperature:Refrigerator
EINECS:206-716-1
Related Information
Frequently Bought Together