Azure B (azure I) ≥89% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:531-55-5
:
Formula:C₁₅H₁₆ClN₃S
MDL Number:MFCD00011935
:305.83 g/mol
:M-Clarity™ quality level = MQ200
EINECS:208-511-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together