Chicken native avidin (from egg)
A2568.0010
:
- Pack type:Glass bottle for solids
- Pk:10 mg
- CAS No.:1405-69-2
- Protein/peptide type:Native
- Source:Egg
- Species:Chicken
- Storage conditions:Freezer
- MDL No.:MFCD00130572
- Protein/peptide name:Avidin