Caspase-3 inhibitor III
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Specifications
- Storage conditions:Refrigerator
- Protein/peptide name:Caspase-3 Inhibitor III
- Sequence:Ac-DEVD-CMK, Ac-ASP-GLU-VAL-ASP-CMK
- Molecular formula:C₂₁H₃₁CIN₄O₁₁
Specifications
Frequently Bought Together