VWR®, Pronase E
Amber bottle, 5 g
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Off-white to light brown powder from Streptomyces griseus.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Amber bottle
- Pk:5 g
- Cat. No.:E629-5G
- Storage conditions:Freezer
- MDL No.:MFCD02253017
- Tested applications:for biotechnology
- Enzyme name:Pronase E
- CAS No.:9036-06-0
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



