ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
Supplier: Serva
Certificates
About this item
CAS Number:2646-71-1
:
Formula:C₂₁H₂₆N₇Na₄O₁₇P₃
MDL Number:MFCD00036263
:833.35 g/mol
EINECS:220-163-3
Frequently Bought Together