Order Entry
Denmark
Orders LinkContactUsLinkComponent
Glycine, PlusOne™

Glycine, PlusOne™

Glass bottle for solids, 500 g
Catalog # 17-1323-01
Supplier:   Cytiva
Glycine, PlusOne™
Catalog # 17-1323-01
Supplier:  Cytiva
CAS Number:  56-40-6
Glycine, PlusOne™
Glycine, PlusOne™

Glass bottle for solids, 500 g

CAS Number:  56-40-6
Price
667,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Glycine is frequently used as a component of electrophoresis buffers, especially in the Laemmli discontinuous buffer system for SDS-PAGE.

  • Soluble in cold water and hot water
  • Slightly soluble in acetone, insoluble in diethyl ether
Synonyms:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS Number:56-40-6
  • Formula:NH₂CH₂COOH
  • MDL Number:MFCD00008131
  • Molecular Weight:75.07 g/mol
  • Merck Index:13,04504
  • Boiling Pt:181 °C (1013 hPa)
  • Melting Pt:245 °C
  • Density:1.16 g/cm³
  • Storage Temperature:Ambient
  • EINECS:200-272-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.