Order Entry
Denmark
ContactUsLinkComponent
 
Gadolinium standard solution, 1,000 mg/L Gd in dilute nitric acid (de Gd2O3) standard for AAS and ICP
  PERCN9300118
 :  
Gadolinium standard solution, 1,000 mg/L Gd in dilute nitric acid (de Gd2O3) standard for AAS and ICP
  PERCN9300118
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Gadolinium
  • Symbol:
    Gd