Corning® Human Recombinant Fibroblast Growth Factors (from E. coli)
Catalog # 734-1405
Supplier: Corning
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 14001 - Bedford
- Protein synonyms:Basic Fibroblast Growth Factor
- Protein/peptide name:bFGF
- Purity:Purity > 97%by SDS-PAGE
Specifications
About this item
Basic Fibroblast Growth Factors (bFGF), human recombinant, are heparin-binding mitogenic proteins that enhance proliferation of a wide variety of cell types under serum-free or serum-reduced conditions.
Can also be used as a chemotactic or neurotrophic factor and to study wound healing, angiogenesis, and related processes.
Tested for ability to promote proliferation of fetal bovine heart endothelial cells (FBHEC).