Order Entry
Denmark
Orders LinkContactUsLinkComponent
KPG® Stirrer Bearings
KPG® Stirrer Bearings
Stirrer bearings, KPG®, interchangeable, ground, HA 10
Catalog # SCOT286304002
Supplier:   DWK Life Sciences
KPG® Stirrer Bearings
Catalog # SCOT286304002
Supplier:  DWK Life Sciences
KPG® Stirrer Bearings
KPG® Stirrer Bearings
Stirrer bearings, KPG®, interchangeable, ground, HA 10
Price
177,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, clear borosilicate glass 3.3
DURAN® KPG® bearing sleeves are used as components for agitator guides/agitator coolers. They are available as ground or polished versions in the following designs:
HA: without grease cup
HB: with grease cup
Specifications
  • For:
    HA 10
  • Description:
    Stirrer bearings, KPG®, interchangeable, ground
  • Seq:
    02