Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spigot, brass
Spigot, brass
3/4" outer
Catalog # BURK0535-0010
Supplier:   BURKLE
Spigot, brass
Catalog # BURK0535-0010
Supplier:  BURKLE
Spigot, brass
Spigot, brass
3/4" outer
Price
506,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Barrel tap for petrochemical liquids.
  • Seals NBR
  • NW 10 mm
  • Closed independently when not used (increased safety)
Specifications
  • Thread:
    3/4" outer