Absorbent skimmer socks, PIG®
Supplier: NEW PIG
Jump to:
About this item
Skimmer socks are used to absorb oil from acid baths. Chemical-resistant PP skin and filler pulls oils away from acids, chemicals, rainwater and other water-based liquids.
- Easy to retrieve — floats on surface even after fully saturated
- Skin remains UV resistant for up to 12 months
- Attached string allows socks to be tied down, tied together or dipped into tanks
Certifications: Meets NFPA 99 standards.
2 Options Available Below
Product Details & Documents
Skimmer socks are used to absorb oil from acid baths. Chemical-resistant PP skin and filler pulls oils away from acids, chemicals, rainwater and other water-based liquids.
- Easy to retrieve — floats on surface even after fully saturated
- Skin remains UV resistant for up to 12 months
- Attached string allows socks to be tied down, tied together or dipped into tanks
Certifications: Meets NFPA 99 standards.
Product Family Options
Product Information
- AbsorbencyPackedØ×LAvailabilityPrice
- NEWP104PSPIG® SKIMMER socks, repel water based liquidsPIG® SKIMMER socks, repel water based liquids38 l/box10 socks/box80×1220 mmLog in for availability1 * 10 Itemskr1,050.00
- NEWPSKM203PIG® SKIMMER socks, repel water based liquidsPIG® SKIMMER socks, repel water based liquids57 l/box6 socks/box80×3000 mmLog in for availability1 * 6 Itemskr1,220.00