Order Entry
Denmark
Orders LinkContactUsLinkComponent
Safety funnels
Safety funnels
Certificates
Safety funnels
16 Options Available Below
Product Details & Documents
Stainless steel.
  • Safety funnels need to be screwed to the container - offer overfill protection, flame trap, grounding, earth connection and closure head
  • Removable flame traps for easy cleaning
  • Safety funnels with plastic cap: Safe and comfortable filling of plastic containers (e.g. for waste solvents)
  • Safety funnels with buttress threads 2" (S56×4): Safe filling of combi-barrels
  • Standard edition I: Ø 120 mm, height above thread 140 mm
  • Flat edition II: Ø 180 mm, height above thread 70 mm
  • Plug-in funnels with Ø 120 mm, without closure head
Funnels allow for safe and comfortable filling of safety and other containers (e.g. plastic canisters).
Product Family Options
Product Information
  • Availability
    Price
  • 119-0152
    Plug-in funnel for all safety barrels
    Plug-in funnel for all safety barrels
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 567,00 kr
    567,00 kr
    1 * 1 Items
       567,00 kr
  • 119-0151
    Plug-in funnel for all safety canisters, with overfill protection
    Plug-in funnel for all safety canisters, with overfill protection
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.040,00 kr
    1.040,00 kr
    1 * 1 Items
       1.040,00 kr
  • 119-0134
    Plug-in funnel inclined for all safety canisters, TR 2
    Plug-in funnel inclined for all safety canisters, TR 2
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 591,00 kr
    591,00 kr
    1 * 1 Items
       591,00 kr
  • 119-2410
    Plug-in funnel straight for all safety cans, TR 1
    Plug-in funnel straight for all safety cans, TR 1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 534,00 kr
    534,00 kr
    1 * 1 Items
       534,00 kr
  • 119-0155
    Safety funnel with buttress thread 2" (S56×4) (II)
    Safety funnel with buttress thread 2" (S56×4) (II)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.290,00 kr
    2.290,00 kr
    1 * 1 Items
       2.290,00 kr
  • 119-0153
    Safety funnel with buttress-thread 2" (S56×4) (I)
    Safety funnel with buttress-thread 2" (S56×4) (I)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.530,00 kr
    1.530,00 kr
    1 * 1 Items
       1.530,00 kr
  • 119-0145
    Safety funnel with plastic cap (PE), DIN 50 (I)
    Safety funnel with plastic cap (PE), DIN 50 (I)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.510,00 kr
    1.510,00 kr
    1 * 1 Items
       1.510,00 kr
  • 119-0146
    Safety funnel with plastic cap (PE), DIN 50 (II)
    Safety funnel with plastic cap (PE), DIN 50 (II)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.380,00 kr
    2.380,00 kr
    1 * 1 Items
       2.380,00 kr
  • 119-0132
    Safety funnel with plastic cap (PE), DIN 51 (I)
    Safety funnel with plastic cap (PE), DIN 51 (I)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.510,00 kr
    1.510,00 kr
    1 * 1 Items
       1.510,00 kr
  • 119-0147
    Safety funnel with plastic cap (PE), DIN 51 (II)
    Safety funnel with plastic cap (PE), DIN 51 (II)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.380,00 kr
    2.380,00 kr
    1 * 1 Items
       2.380,00 kr
  • 119-0133
    Safety funnel with plastic cap (PE), DIN 61 (I)
    Safety funnel with plastic cap (PE), DIN 61 (I)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.510,00 kr
    1.510,00 kr
    1 * 1 Items
       1.510,00 kr
  • 119-0148
    Safety funnel with plastic cap (PE), DIN 61 (II)
    Safety funnel with plastic cap (PE), DIN 61 (II)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.260,00 kr
    2.260,00 kr
    1 * 1 Items
       2.260,00 kr
  • 119-0149
    Safety funnel with plastic cap (PE), DIN 71 (I)
    Safety funnel with plastic cap (PE), DIN 71 (I)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
  • 119-0150
    Safety funnel with plastic cap (PE), DIN 71 (II)
    Safety funnel with plastic cap (PE), DIN 71 (II)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.290,00 kr
    2.290,00 kr
    1 * 1 Items
       2.290,00 kr
  • 119-0131
    Safety funnel with thread 2" (I)
    Safety funnel with thread 2" (I)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.520,00 kr
    1.520,00 kr
    1 * 1 Items
       1.520,00 kr
  • 119-0141
    Safety funnel with thread 2" (II)
    Safety funnel with thread 2" (II)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.290,00 kr
    2.290,00 kr
    1 * 1 Items
       2.290,00 kr