Portable photometers, MultiDirect
LOVI210000
: Lovibond
- Light source:LED
- Description:MultiDirect Photometer
- Seq:01
- Model:MultiDirect
- Display:Graphical
- Interfaces:RS232 for printer and PC-connection
- Keypad:Acid and solvent resistant, touch-sensitive keypad
- Operating conditions:5 - 40 °C; up to 90% RH
- Optics:6 temperature compensating LEDs, internal reference channel, photodiode in protected sample chamber
- Power supply:7 Ni-MH-battery pack supply (AA/Mignon), charged whilst in the unit with external mains charger, integrated overload cut-out
- Storage capacity:1000 data sets with date, time and registration number
- System check:Auto-check and Auto-off
- Wavelengths:430, 530, 560, 580, 610, 660 nm
- Weight:1 kg
- W×D×H:265×195×70 mm
- Pk:1
The MultiDirect is a microprocessor controlled photometer with ergonomically designed keypad and large format graphic display. It is equipped with preprogrammed methods based on Lovibond® tablet, liquid and powder reagents.
- High accuracy from dual beam technology
- Instrument and sample chamber is waterproof
- Long term stable LED light sources
- Update of new methods and languages via the internet
: Supplied with rechargeable batteries, mains charger, PC connection cable, carrying case, 3× round vials 24 mm, 3× round vials 16 mm, 16 mm vial adapter, 3×syringes and 100 ml beaker.