0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Closures
Bottle Closures
Pouring rings for laboratory bottles
Pouring rings for laboratory bottles
WITG0.175.201
:
witeg Labortechnik
Pouring rings for laboratory bottles
WITG0.175.201
:
witeg Labortechnik
Thread:
GL 32
Colour:
Blue
Pk:
1
Pouring rings, made of polypropylene.
For DIN GL threads
Heat resistant up to +140 °C
Good chemical resistance