Order Entry
Denmark
Orders LinkContactUsLinkComponent
Standard ground joints, double, cone
Standard ground joints, double, cone
Leverandør:   witeg Labortechnik
Standard ground joints, double, cone
Leverandør:  witeg Labortechnik
Certifikater
Standard ground joints, double, cone
Standard ground joints, double, cone
2 tilgængelige muligheder nedenfor
Product Details & Documents
Made of borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Certifications: DIN 12 249.
Product Family Options
Produktinformation
  • Cone size
    Total length
    Tube ext.Ø
    Wall thickness
    Tilgængelighed
    Price
  • 24/29 NS
    170 mm
    22 mm
    1,8 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 133,00 kr
    133,00 kr
    1 * 1 Items
       133,00 kr
  • 29/32 NS
    170 mm
    26 mm
    2,0 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 134,00 kr
    134,00 kr
    1 * 1 Items
       134,00 kr