Human alpha-chymotrypsin (from Pancreas)
Catalog # 230900-50
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:50 µG
- CAS No.:9004-07-3
- Source:Pancreas
- Species:Human
- Storage conditions:Freezer
- MDL No.:MFCD00130481
- Enzyme name:alpha-Chymotrypsin