Order Entry
Denmark
ContactUsLinkComponent
Polyethylene glycol dimethylether, M.W. 500 stabilised for synthesis, Sigma-Aldrich®
Polyethylene glycol dimethylether, M.W. 500 stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.14171.0500
Supplier:  Merck
CAS Number:  
Polyethylene glycol dimethylether, M.W. 500 stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.14171.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 ml
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    13 °C
  • Molar Mass:
    530.65 g/mol
  • Boiling Point:
    250 °C (1013 hPa)
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    220 °C
  • Refractive Index:
    1.454 (20 °C, 589 nm)
  • Density:
    1.07 g/cm3 (20 °C)
  • pH:
    (100 g/l, H2O, 20 °C) neutral
  • Ignition Point:
    360 °C
  • Viscosity kinematic:
    15 mm2/s (40 °C)

Specification Test Results

About this item

Synonyms: Poly(ethylene glycol) dimethyl ether Mn 250
 :
 :M-Clarity™ quality level = MQ200
CAS Number:24991-55-7
Formula:H₃C(OCH₂CH₂)nOCH₃
MDL Number:MFCD00147972