Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Workwear
Fristads Latzhose PR54-22 Marine Design B
Fristads Latzhose PR54-22 Marine Design B
Fristads Latzhose PR54-22 Marine Design B
Catalog # KWINPR5422B1552
Supplier:
FRISTADS KANSAS
Expand image
Fristads Latzhose PR54-22 Marine Design B
Catalog # KWINPR5422B1552
Supplier:
FRISTADS KANSAS
Specifications
Garment fit:
Regular
Garment application:
General Purpose
,
Industrial
Colour:
Green
Pk:
1
Specifications
About this item
Product Details
Documentation
Product Details
Documentation