Miller® DuraFlex® AGU 90S Elasticated Harness
Catalog # 139-0243
Supplier: HONEYWELL SAFETY
Specifications
- Size:M/L
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Four point safety harness with integrated positioning belt, rear fall arrest eyelet and 2 retaining eyelets.
- Made from DuraFlex® (highly elastic stretch fabric): adapts to the user's body shape and expands with every movement so that the straps do not dig in, hinder or get twisted
- Additionally treated with PTFE
- Additional arrester eyelet, all eyelets made from stainless steel
- Quick-release closures
- Unrivalled comfort and freedom of movement as well as a soft catch in the event of a fall
Certifications: EN 361.