Order Entry
Denmark
ContactUsLinkComponent
Tetracycline hydrochloride, Supelco®
Irritant
Tetracycline hydrochloride, Supelco®
Catalog # 31741-250MG
Supplier:  Merck
CAS Number:  
undefined
Irritant
Tetracycline hydrochloride, Supelco®
Catalog # 31741-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :480.9 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:200-593-8
 :GHS07
Storage Temperature:Ambient
CAS Number:64-75-5
Formula:C₂₂H₂₅ClN₂O₈
MDL Number:MFCD00078142