
Tetracycline hydrochloride
Catalog # PROOICRS0429
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:200 mg
- Seq:2530
Specifications
About this item
:
:480.9 g/mol
EINECS:200-593-8
:GHS07
Storage Temperature:Ambient
CAS Number:64-75-5
Formula:C₂₂H₂₅ClN₂O₈
MDL Number:MFCD00078142