

Tetracycline hydrochloride 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64-75-5
:
Formula:C₂₂H₂₅ClN₂O₈
MDL Number:MFCD00078142
:480.9 g/mol
Merck Index:14,9196
Melting Pt:220…223 °C
EINECS:200-593-8
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together