Order Entry
Denmark
ContactUsLinkComponent
 
Bovine Ribonuclease A (from Pancreas)
  55674-50
 :  
Bovine Ribonuclease A (from Pancreas)
  55674-50
 :  

 

  • Pk:
    50 KU
  • CAS No.:
    9001-99-4
  • Source:
    Pancreas
  • Species:
    Bovine
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00132184
  • Enzyme name:
    Ribonuclease A
  • Enzyme synonyms:
    RNase A