Ampicillin
Glass bottle for solids, 200 mg
Catalog # PROOICRS0211
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:69-53-4
- Formula:C₁₆H₁₉N₃O₄S
- MDL Number:MFCD00005175
- Molecular Weight:349.4 g/mol
- EINECS:200-709-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:200 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



