Test tube racks, Origami TubeRack
: TENAK
PP
- Height adjustable which makes it very flexible for a wide range of tubes
- Alphanumeric grid for easy location of the samples
- Can be used to store samples from –45 to 60 °C
An innovative design that is delivered flat packed to reduce shipping cost and storage space. Easy to assemble.
Frequently Bought Together