Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Abietic acid 85%

Irritant
Environment
Abietic acid 85%
Certificates
Abietic acid 85%

2 Options Available Below

Product Details & Documents
  • CAS Number:514-10-3
  • Formula:C₂₀H₃₀O₂
  • MDL Number:MFCD03423567
  • Molecular Weight:302.45 g/mol
  • Storage Temperature:Ambient
  • EINECS:208-178-3
  • UN:3077
  • ADR:9,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • GC:
    ≥80 %
  • Free fructose, glucose,sucrose:
    ≤5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 536,00 kr
    536,00 kr
    1 * 25 g
       536,00 kr

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1.420,00 kr
    1.420,00 kr
    1 * 100 g
       1.420,00 kr

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.