
(S)-(+)-Prolinol 95%
:23356-96-9
:
:C₅H₁₁NO
:MFCD00005255
:101.15 g/mol
:74…76 °C (2 mmHg)
:1.036 g/cm³
:86 °C
:245-605-2
:GHS07
:Refrigerator
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4840 to 1.4870 (20°C, 589 nm)
- GC:≥94 %
- Specific optical rotation:+29° to +33° (20°C, 589 nm) (c=1, toluene)
Frequently Bought Together